Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 8 | 1746 | ||
NM_024860.3 | NP_079136.2 | 9 | 1674 | |||
NR_134583.1 | 9 | 1705 | ||||
GenBank mRNA | AK024801.1 | - | 9 | 1630 | ||
AK127136.1 | - | 5 | 3151 | |||
BC110433.1 | - | 9 | 1689 |