Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098517.1 | NP_001091987.1 | 9 | 2266 | ||
NM_001301043.1 | NP_001287972.1 | 12 | 2437 | |||
NM_001301044.1 | NP_001287973.1 | 11 | 2383 | |||
NM_001301045.1 | NP_001287974.1 | 11 | 2353 | |||
NM_014333.3 | NP_055148.3 | 10 | 2350 | |||
XM_005271494.2 | XP_005271551.1 | 10 | 2192 | |||
XM_017017457.1 | XP_016872946.1 | 12 | 2396 | |||
XM_017017458.1 | XP_016872947.1 | 10 | 2116 | |||
XM_017017459.1 | XP_016872948.1 | 13 | 4390 | |||
XM_017017460.1 | XP_016872949.1 | 11 | 4303 | |||
XM_017017461.1 | XP_016872950.1 | 10 | 2233 | |||
GenBank mRNA | AF132811.1 | - | 10 | 2226 | ||
BC125102.1 | - | 9 | 2141 | |||
BC125103.1 | - | 9 | 2140 | 40118330 |