Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 19 | 3086 | ||
NM_182931.2 | NP_891847.1 | 20 | 3234 | |||
XM_005250493.1 | XP_005250550.1 | 19 | 3153 | |||
XM_011516400.1 | XP_011514702.1 | 21 | 3360 | |||
XM_017012435.1 | XP_016867924.1 | 20 | 3234 | |||
GenBank mRNA | AF519459.1 | - | 19 | 2901 | ||
AY147037.1 | - | 20 | 3234 | |||
AY157990.1 | - | 17 | 2460 | |||
JQ809698.1 | - | 18 | 2700 |