Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145527.1 | NP_001138999.1 | 5 | 977 | ||
NM_024757.4 | NP_079033.4 | 5 | 977 | |||
XM_005266105.4 | XP_005266162.1 | 5 | 1020 | |||
XM_005266110.1 | XP_005266167.1 | 4 | 892 | |||
XM_006717288.2 | XP_006717351.1 | 5 | 938 | |||
XM_011519021.2 | XP_011517323.1 | 5 | 1975 | |||
XM_011519022.2 | XP_011517324.1 | 5 | 1430 | |||
XM_011519023.2 | XP_011517325.1 | 5 | 1954 | |||
XM_011519024.2 | XP_011517326.1 | 5 | 1976 | |||
XM_011519025.1 | XP_011517327.1 | 5 | 1140 | |||
XM_011519026.2 | XP_011517328.1 | 5 | 1977 | |||
XM_011519027.2 | XP_011517329.1 | 5 | 1976 | |||
XM_011519028.2 | XP_011517330.1 | 5 | 1976 | |||
XM_011519033.2 | XP_011517335.1 | 5 | 1956 | |||
XM_017015134.1 | XP_016870623.1 | 5 | 1408 | |||
XM_017015135.1 | XP_016870624.1 | 5 | 960 | |||
XM_017015136.1 | XP_016870625.1 | 5 | 960 | |||
XM_017015137.1 | XP_016870626.1 | 5 | 1119 | |||
XM_017015138.1 | XP_016870627.1 | 4 | 871 | |||
GenBank mRNA | AK022941.1 | - | 4 | 892 | ||
AK309602.1 | - | 2 | 295 | |||
AY083210.1 | - | 3 | 847 | |||
BC025772.1 | - | 5 | 967 | |||
BC047504.1 | - | 5 | 977 | 5298029 |