Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT2C SIRNA Assays ›
Gene Symbol:
KMT2C
Gene Name:
lysine methyltransferase 2C
Gene Aliases:
HALR, MLL3
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
UniGene ID:
Hs.647120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
214048
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_170606.2
NP_733751.2
34
5280
XM_005250025.4
XP_005250082.1
36
5134
XM_005250026.3
XP_005250083.1
36
5168
XM_005250027.4
XP_005250084.1
35
5342
XM_005250028.4
XP_005250085.1
35
5342
XM_005250031.4
XP_005250088.1
35
5342
XM_006716077.3
XP_006716140.1
35
5342
XM_006716078.3
XP_006716141.1
35
5342
XM_006716079.3
XP_006716142.1
35
5342
XM_011516450.2
XP_011514752.1
34
5294
XM_011516451.2
XP_011514753.1
34
5222
XM_011516452.2
XP_011514754.1
33
5189
XM_011516453.2
XP_011514755.1
35
5342
XM_011516454.2
XP_011514756.1
30
4337
XM_011516456.2
XP_011514758.1
34
5294
XM_017012480.1
XP_016867969.1
36
5171
XM_017012481.1
XP_016867970.1
36
5131
XM_017012482.1
XP_016867971.1
35
5342
XM_017012483.1
XP_016867972.1
35
5342
XM_017012484.1
XP_016867973.1
35
5309
XM_017012485.1
XP_016867974.1
34
5291
XM_017012486.1
XP_016867975.1
35
5342
XM_017012487.1
XP_016867976.1
35
5310
XM_017012488.1
XP_016867977.1
35
5342
XM_017012489.1
XP_016867978.1
16
1849
XM_017012490.1
XP_016867979.1
13
1421
GenBank mRNA
AB040939.2
-
23
3396
AF264750.2
-
20
2607
AY024361.1
-
34
5241
BC151837.1
-
23
3396
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0