Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193424.1 | NP_001180353.1 | 6 | 1719 | ||
NM_001193425.1 | NP_001180354.1 | 6 | 1677 | |||
NM_001193426.1 | NP_001180355.1 | 6 | 1179 | |||
NM_001193427.1 | NP_001180356.1 | 6 | 1137 | |||
NM_024670.3 | NP_078946.1 | 5 | 1664 | |||
NR_034181.1 | 5 | 1573 | ||||
XM_006717503.3 | XP_006717566.1 | 6 | 1920 | |||
XM_011519662.2 | XP_011517964.1 | 6 | 1708 | |||
XM_017016637.1 | XP_016872126.1 | 5 | 1614 | |||
GenBank mRNA | AK027067.1 | - | 5 | 1470 | ||
AK222565.1 | - | 5 | 1480 | |||
AK225369.1 | - | 5 | 1472 | |||
AL834488.1 | - | 4 | 1328 | |||
BC007754.2 | - | 5 | 1664 | 4109807 | ||
BC029360.1 | - | 6 | 1099 | 4079431 |