Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 6 | 1267 | ||
NM_139003.2 | NP_620572.1 | 5 | 949 | |||
NM_139004.2 | NP_620573.1 | 5 | 991 | |||
NM_139006.2 | NP_620575.1 | 6 | 1225 | |||
NM_139007.2 | NP_620576.1 | 5 | 1003 | |||
NM_139008.2 | NP_620577.1 | 5 | 961 | |||
NM_139009.2 | NP_620578.1 | 6 | 1198 | |||
NM_139010.2 | NP_620579.1 | 4 | 727 | |||
NM_139011.2 | NP_620580.1 | 3 | 451 | |||
GenBank mRNA | AF115265.1 | - | 6 | 1118 | ||
AF144242.1 | - | 5 | 954 | |||
AJ249335.1 | - | 6 | 1198 | |||
AJ249336.1 | - | 5 | 1003 | |||
AJ249337.1 | - | 5 | 991 | |||
AJ249338.1 | - | 3 | 451 | |||
AJ250635.1 | - | 4 | 727 | |||
BC117201.1 | - | 6 | 1142 | 40125752 | ||
BC117203.1 | - | 6 | 1142 | 40125754 | ||
U60319.1 | - | 6 | 1328 |