Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003064.1 | XP_016858553.1 | 29, 30 | 4823 | ||
GenBank mRNA | CR749273.1 | - | 9, 10 | 2091 |