Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 2, 3 | 268 | ||
GenBank mRNA | AI302038.1 | - | 2, 3 | 169 | ||
BC007783.2 | - | 2, 3 | 263 | 4308472 | ||
BC146919.1 | - | 2, 3 | 229 | 9056847 | ||
BC147008.1 | - | 2, 3 | 229 | 9056936 | ||
BC147013.1 | - | 2, 3 | 229 | 9056941 | ||
BT006783.1 | - | 2, 3 | 193 | |||
CF128666.1 | - | 2, 3 | 247 |
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D63785.1 | - | 2, 3 | 173 |
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 2, 3 | 278 | ||
NR_111964.1 | 2, 3 | 470 | ||||
GenBank mRNA | AV739733.1 | - | 2, 3 | 322 | ||
BC027888.1 | - | 2, 3 | 266 | 5222247 | ||
BC107710.1 | - | 2, 3 | 282 | 6699133 | ||
BC146914.1 | - | 2, 3 | 229 | 9056842 | ||
X52149.1 | - | 2, 3 | 456 |