Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007160.3 | NP_009091.3 | 1 | 560 | ||
XM_011514912.2 | XP_011513214.1 | 1 | 1942 | |||
GenBank mRNA | BC069146.1 | - | 1 | 521 | 7217006 | |
BC101683.1 | - | 1 | 560 | 8069189 | ||
BC112176.1 | - | 1 | 572 | 8327644 |