Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278241.1 | NP_001265170.1 | 17 | 3061 | ||
NM_014389.2 | NP_055204.3 | 16 | 2979 | |||
GenBank mRNA | AF547989.1 | - | 16 | 2753 | ||
AK226056.1 | - | 16 | 2759 | |||
AK291163.1 | - | 16 | 2779 | |||
AK293838.1 | - | 17 | 2848 | |||
AK299084.1 | - | 16 | 2967 | |||
AY882602.1 | - | 16 | 2546 | |||
BC002875.2 | - | 6 | 1594 | |||
BC010457.2 | - | 15 | 2507 | |||
BC069058.1 | - | 16 | 2753 | 6068755 | ||
U88153.2 | - | 15 | 3215 | |||
U88154.1 | - | 15 | 2426 |