Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002253.2 | NP_002244.1 | 30 | 4365 | ||
GenBank mRNA | AB209901.1 | - | 30 | 4350 | ||
AF035121.1 | - | 30 | 4366 | |||
AK293668.1 | - | 4 | 527 | |||
BC131822.1 | - | 30 | 4125 | 40129287 | ||
L04947.1 | - | 30 | 4057 | |||
X61656.1 | - | 30 | 4057 |