Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 24 | 3462 | ||
XM_005259896.3 | XP_005259953.1 | 24 | 3501 | |||
GenBank mRNA | BM561964.1 | - | 1 | 170 | ||
U09607.1 | - | 23 | 3457 | |||
U31317.1 | - | 1 | 155 |