Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF128210.1 | - | 5, 6 | 588 | ||
AF128211.1 | - | 5, 6 | 588 | |||
AF128212.1 | - | 6, 7 | 515 | |||
AF128213.1 | - | 6, 7 | 515 | |||
AK153729.1 | - | 2, 3 | 645 | |||
BC116851.1 | - | 6, 7 | 519 | |||
BC116855.1 | - | 6, 7 | 519 | |||
M86672.1 | - | 6, 7 | 617 |