Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 15 | 1308 | ||
GenBank mRNA | AF074881.1 | - | 15 | 1255 | ||
AF074882.1 | - | 9 | 682 | |||
AF098295.1 | - | 15 | 1233 | |||
AF125536.1 | - | 15 | 1240 | |||
AK037478.1 | - | 1300 | ||||
AK145158.1 | - | 15 | 1274 | |||
AK189827.1 | - | 2 | 77 | |||
BC139300.1 | - | 15 | 1336 | 8862322 | ||
BC139301.1 | - | 15 | 1336 | 8862323 | ||
JN651901.1 | - | 9 | 956 | |||
JN651902.1 | - | 8 | 914 | |||
JN651903.1 | - | 9 | 665 | |||
JN651904.1 | - | 6 | 510 |