Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322227.1 | NP_001309156.1 | 14 | 1925 | ||
NM_001322228.1 | NP_001309157.1 | 12 | 1657 | |||
NM_001322229.1 | NP_001309158.1 | 13 | 1765 | |||
NM_001322230.1 | NP_001309159.1 | 14 | 1952 | |||
NM_001322231.1 | NP_001309160.1 | 12 | 1661 | |||
NM_001322232.1 | NP_001309161.1 | 12 | 1546 | |||
NM_001322233.1 | NP_001309162.1 | 12 | 1603 | |||
NM_001322234.1 | NP_001309163.1 | 13 | 1802 | |||
NM_001322235.1 | NP_001309164.1 | 14 | 1905 | |||
NM_001322236.1 | NP_001309165.1 | 12 | 1681 | |||
NM_001322237.1 | NP_001309166.1 | 13 | 1564 | |||
NM_001322238.1 | NP_001309167.1 | 11 | 1273 | |||
NM_004755.3 | NP_004746.2 | 13 | 1765 | |||
NM_182398.2 | NP_872198.1 | 13 | 1765 | |||
XM_017021786.1 | XP_016877275.1 | 14 | 1804 | |||
GenBank mRNA | AF074393.1 | - | 13 | 1715 | ||
AF080000.1 | - | 13 | 1731 | |||
AF090421.1 | - | 13 | 1714 | |||
AK295266.1 | - | 13 | 1564 | |||
BC017187.2 | - | 13 | 1716 | 3139114 |