Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003188.3 | NP_003179.1 | 7 | 1108 | ||
NM_145331.2 | NP_663304.1 | 7 | 1108 | |||
NM_145332.2 | NP_663305.1 | 7 | 1108 | |||
NM_145333.2 | NP_663306.1 | 7 | 1108 | |||
XM_006715553.2 | XP_006715616.1 | 7 | 896 | |||
XM_017011226.1 | XP_016866715.1 | 7 | 896 | |||
GenBank mRNA | AB009356.1 | - | 7 | 852 | ||
AB009357.1 | - | 7 | 852 | |||
AB009358.2 | - | 7 | 690 | |||
AF218074.1 | - | 7 | 696 | |||
AK299268.1 | - | 5 | 656 | |||
AK315774.1 | - | 7 | 876 | |||
BC017715.2 | - | 7 | 875 | 3906837 | ||
BT019654.1 | - | 7 | 690 | |||
BT019655.1 | - | 7 | 690 |