Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271957.1 | NP_001258886.1 | 4 | 1609 | ||
NM_001271958.1 | NP_001258887.1 | 4 | 1446 | |||
NM_001271959.1 | NP_001258888.1 | 4 | 1615 | |||
NM_001271960.1 | NP_001258889.1 | 5 | 1507 | |||
NM_001271961.1 | NP_001258890.1 | 3 | 1315 | |||
NM_014437.4 | NP_055252.2 | 5 | 1753 | |||
GenBank mRNA | AF132942.1 | - | 3 | 1303 | ||
AF151829.1 | - | 4 | 1324 | |||
AJ243649.1 | - | 4 | 1340 | |||
AJ271671.1 | - | 4 | 1355 | |||
AK074943.1 | - | 4 | 1563 | |||
AK075257.1 | - | 4 | 1377 | |||
AK293389.1 | - | 3 | 1223 | |||
BC002563.2 | - | 4 | 1328 | 3138575 | ||
BC003152.2 | - | 3 | 1300 | 2958651 | ||
BC007886.2 | - | 4 | 1325 | 4122086 | ||
BC014303.2 | - | 5 | 1733 | 3942586 | ||
BM807264.1 | - | 1 | 73 |