Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012303.2 | NP_001012303.1 | 5 | 827 | ||
NM_006217.4 | NP_006208.1 | 4 | 736 | |||
GenBank mRNA | AB006423.1 | - | 4 | 653 | ||
AF130470.1 | - | 3 | 662 | |||
AK226025.1 | - | 4 | 626 | |||
AK293391.1 | - | 4 | 652 | |||
BC027859.2 | - | 4 | 697 | 5228038 |