Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000528.3 | NP_000519.2 | 14 | 1811 | ||
NM_001173498.1 | NP_001166969.1 | 14 | 1808 | |||
XM_005259913.1 | XP_005259970.1 | 14 | 1792 | |||
XM_017026818.1 | XP_016882307.1 | 11 | 1390 | |||
GenBank mRNA | AB209921.1 | - | 14 | 1723 | ||
AK291572.1 | - | 14 | 1776 | |||
AK303418.1 | - | 15 | 1750 | |||
BC000736.2 | - | 14 | 1757 | 3504417 | ||
U05572.1 | - | 14 | 1765 | |||
U60266.1 | - | 14 | 1749 | |||
U68382.1 | - | 12 | 2324 | |||
U68567.1 | - | 14 | 1723 |