Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001895.2 | NP_001001895.1 | 12, 13 | 1717 | ||
NM_018961.3 | NP_061834.1 | 13, 14 | 1831 | |||
XM_006724013.3 | XP_006724076.1 | 13, 14 | 1915 | |||
XM_011529606.2 | XP_011527908.1 | 13, 14 | 1800 | |||
XM_011529607.2 | XP_011527909.1 | 13, 14 | 1884 | |||
XM_011529610.2 | XP_011527912.1 | 9, 10 | 3830 | |||
GenBank mRNA | AF520809.1 | - | 13, 14 | 1758 | ||
AF521702.1 | - | 12, 13 | 1644 | |||
AJ277750.1 | - | 13, 14 | 1778 | |||
AK308151.1 | - | 12, 13 | 1658 | |||
AK313367.1 | - | 12, 13 | 1856 | |||
BC028138.1 | - | 12, 13 | 2185 | 5215324 | ||
BC069357.1 | - | 13, 14 | 1778 | 7262315 | ||
BC069483.1 | - | 13, 14 | 1778 | 7262391 | ||
BC069511.1 | - | 12, 13 | 1662 | 7262403 |