Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002214.2 | NP_002205.1 | 12 | 2648 | ||
XM_011515393.2 | XP_011513695.1 | 13 | 2451 | |||
XM_011515394.2 | XP_011513696.1 | 13 | 2049 | |||
XM_011515396.1 | XP_011513698.1 | 13 | 2720 | |||
XM_017012178.1 | XP_016867667.1 | 13 | 2345 | |||
XM_017012179.1 | XP_016867668.1 | 13 | 2804 | |||
XM_017012180.1 | XP_016867669.1 | 13 | 3097 | |||
XM_017012181.1 | XP_016867670.1 | 14 | 2916 | |||
XM_017012182.1 | XP_016867671.1 | 13 | 2031 | |||
XM_017012183.1 | XP_016867672.1 | 13 | 1988 | |||
GenBank mRNA | AK295044.1 | - | 13 | 2720 | ||
M73780.1 | - | 12 | 2623 |