Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302777.1 | NP_001289706.1 | 5 | 1111 | ||
NM_004332.3 | NP_004323.2 | 4 | 883 | |||
NR_026648.2 | 5 | 1111 | ||||
NR_026649.2 | 4 | 883 | ||||
GenBank mRNA | AJ617684.1 | - | 4 | 531 | ||
AK297518.1 | - | 4 | 567 | |||
AK314909.1 | - | 5 | 765 | |||
AL832502.1 | - | 4 | 534 | |||
BC037778.1 | - | 5 | 1116 | 4799692 | ||
BC106901.1 | - | 4 | 598 | 40031474 | ||
X81372.1 | - | 5 | 679 |