Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 7 | 1384 | ||
GenBank mRNA | AF251707.1 | - | 7 | 1256 | ||
AK027334.1 | - | 7 | 1173 | |||
AK130144.1 | - | 5 | 681 | |||
BC063431.1 | - | 7 | 1274 | 5229539 |