Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001947.3 | NP_001938.2 | 3 | 1324 | ||
GenBank mRNA | AF508727.1 | - | 3 | 1092 | ||
AF508728.1 | - | 1 | 540 | |||
BC019107.2 | - | 3 | 956 | |||
BC104880.1 | - | 3 | 1152 | 8143883 | ||
BC104882.1 | - | 3 | 1152 | 8143885 | ||
X93921.1 | - | 3 | 954 |