Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040152.1 | NP_001035242.1 | 2 | 813 | ||
NM_001172437.2 | NP_001165908.1 | 2 | 824 | |||
NM_001172438.2 | NP_001165909.1 | 2 | 824 | |||
NM_001184961.1 | NP_001171890.1 | 2 | 813 | |||
NM_001184962.1 | NP_001171891.1 | 2 | 813 | |||
NM_015068.3 | NP_055883.2 | 2 | 813 | |||
GenBank mRNA | AB028974.1 | - | 1 | 386 | ||
AB049150.1 | - | 2 | 592 | |||
AB049834.1 | - | 1 | 451 | |||
AF216076.1 | - | 1 | 344 | |||
AK299837.1 | - | 779 | ||||
BC050659.2 | - | 2 | 745 | 6197322 |