Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025577.2 | NP_001020748.2 | 1 | 3136 | ||
GenBank mRNA | AK033026.1 | - | 51 | |||
AK079582.1 | - | 1 | 1192 | |||
AK134055.1 | - | 1 | 1305 | |||
AK151743.1 | - | 1 | 1047 | |||
BC034073.1 | - | 1 | 1313 | |||
BC041683.1 | - | 1 | 1313 |