Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284334.1 | NP_001271263.1 | 6, 7 | 588 | ||
NM_014509.4 | NP_055324.2 | 8, 9 | 780 | |||
XM_017028739.1 | XP_016884228.1 | 7, 8 | 725 | |||
GenBank mRNA | AK295223.1 | - | 8, 9 | 737 | ||
AL450314.1 | - | 8, 9 | 723 | |||
AL589866.1 | - | 2, 3 | 81 | |||
AL590118.1 | - | 6, 7 | 429 | |||
AL590120.1 | - | 8, 9 | 621 | |||
BC119650.2 | - | 8, 9 | 664 | 40115836 | ||
BC119651.1 | - | 8, 9 | 664 | 40115837 | ||
BC127635.1 | - | 8, 9 | 665 | 40115838 | ||
BC127636.1 | - | 8, 9 | 665 | 40115841 | ||
CR456455.1 | - | 2, 3 | 87 |