Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001852.3 | NP_001001852.2 | 4 | 980 | ||
GenBank mRNA | AB114795.1 | - | 4 | 962 | ||
AK292005.1 | - | 4 | 628 | |||
BC052239.1 | - | 4 | 561 | |||
BC141855.1 | - | 4 | 593 | 8860375 |