Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000967.3 | NP_000958.1 | 10 | 1280 | ||
NM_001033853.1 | NP_001029025.1 | 10 | 1133 | |||
GenBank mRNA | AB062291.1 | - | 9 | 1217 | ||
AK092695.1 | - | 10 | 1091 | |||
AL832757.1 | - | 1 | 5383 | |||
AY320405.1 | - | 3 | 1359 | |||
BC002408.2 | - | 9 | 1215 | 2961541 | ||
BC004323.1 | - | 8 | 879 | |||
BC006483.1 | - | 10 | 1225 | |||
BC008003.1 | - | 9 | 1217 | 3507481 | ||
BC008492.1 | - | 10 | 1238 | 4251511 | ||
BC012146.1 | - | 10 | 1225 | 4549682 | ||
BC012786.2 | - | 10 | 1225 | 3951917 | ||
BC013674.1 | - | 9 | 1221 | 4152521 | ||
BC013861.1 | - | 9 | 1221 | |||
BC014017.2 | - | 9 | 1220 | 4128023 | ||
BC015032.1 | - | 9 | 1221 | |||
BC015767.1 | - | 9 | 1221 | 4861546 | ||
BC022790.1 | - | 9 | 1126 | |||
BC033296.1 | - | 9 | 1218 | |||
BC036582.1 | - | 10 | 2057 | 4796724 | ||
BC063662.1 | - | 9 | 1225 | 3852576 | ||
BC088373.1 | - | 10 | 1235 | 6195715 | ||
BC107711.1 | - | 10 | 1242 | 6699816 | ||
CR456566.1 | - | 9 | 1224 | |||
M90054.1 | - | 9 | 1198 | |||
X73460.1 | - | 9 | 1218 |