Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001511.3 | NP_001502.1 | 3 | 335 | ||
NR_046035.1 | 3 | 358 | ||||
GenBank mRNA | BC011976.1 | - | 3 | 323 | 3856841 | |
BG121608.1 | - | 3 | 342 | |||
BM554448.1 | - | 3 | 348 | |||
BT006880.1 | - | 3 | 256 | |||
CB112830.1 | - | 3 | 336 | |||
J03561.1 | - | 3 | 309 | |||
X12510.1 | - | 3 | 273 |