Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198586.2 | NP_940988.2 | 1 | 897 | ||
GenBank mRNA | AY324850.1 | - | 1 | 897 | ||
BC103888.1 | - | 1 | 897 | 40004980 | ||
BC103889.1 | - | 1 | 897 | 40004983 | ||
BC103890.1 | - | 1 | 897 | 40004985 |