Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002824.4 | NP_002815.3 | 5 | 609 | ||
XM_011520987.1 | XP_011519289.1 | 5 | 922 | |||
GenBank mRNA | BC017025.1 | - | 5 | 328 | 3900998 | |
BC128229.1 | - | 6 | 276 | |||
BE791815.1 | - | 2 | 59 | |||
BM550529.1 | - | 686 | ||||
M24398.1 | - | 5 | 578 |