Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197107.1 | NP_001184036.1 | 2 | 843 | ||
NM_014575.3 | NP_055390.1 | 2 | 843 | |||
GenBank mRNA | AF145713.1 | - | 2 | 338 | ||
AF145715.1 | - | 2 | 338 | |||
AK309223.1 | - | 2 | 290 | |||
AK314662.1 | - | 2 | 786 | |||
BC036535.2 | - | 2 | 846 | 5263291 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 5 | 678 | ||
NM_001197114.1 | NP_001184043.1 | 4 | 597 | |||
GenBank mRNA | AK096479.1 | - | 5 | 584 | ||
DQ157847.1 | - | 3 | 416 | |||
DQ157848.1 | - | 4 | 497 |