Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267042.2 | NP_001253971.1 | 5, 6 | 738 | ||
NM_014154.4 | NP_054873.2 | 6, 7 | 814 | |||
NM_015396.5 | NP_056211.2 | 6, 7 | 814 | |||
NM_213654.3 | NP_998819.1 | 6, 7 | 868 | |||
XM_006713564.3 | XP_006713627.1 | 5, 6 | 733 | |||
XM_006713565.2 | XP_006713628.1 | 6, 7 | 774 | |||
XM_006713566.3 | XP_006713629.1 | 4, 5 | 628 | |||
XM_006713567.3 | XP_006713630.1 | 5, 6 | 733 | |||
XM_011512632.1 | XP_011510934.1 | 5, 6 | 681 | |||
XM_011512633.1 | XP_011510935.1 | 5, 6 | 735 | |||
XM_017006095.1 | XP_016861584.1 | 6, 7 | 736 | |||
XM_017006096.1 | XP_016861585.1 | 4, 5 | 628 | |||
XM_017006097.1 | XP_016861586.1 | 5, 6 | 682 | |||
XM_017006098.1 | XP_016861587.1 | 4, 5 | 628 | |||
XM_017006099.1 | XP_016861588.1 | 5, 6 | 719 | |||
XM_017006100.1 | XP_016861589.1 | 5, 6 | 733 | |||
GenBank mRNA | AF109184.1 | - | 6, 7 | 567 | ||
AF161541.1 | - | 6, 7 | 536 | |||
AK001721.1 | - | 4, 5 | 376 | |||
AK289577.1 | - | 6, 7 | 510 | |||
AK296757.1 | - | 5, 6 | 469 | |||
BC013424.1 | - | 6, 7 | 568 | 3892143 | ||
BC032661.1 | - | 6, 7 | 678 | 5530425 | ||
BC041699.1 | - | 6, 7 | 493 | 5240423 | ||
CX167429.1 | - | 5, 6 | 443 |