Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213604.2 | NP_998769.2 | 12 | 2375 | ||
XM_005259549.3 | XP_005259606.1 | 13 | 2084 | |||
XM_011527961.1 | XP_011526263.1 | 13 | 2096 | |||
XM_011527963.1 | XP_011526265.1 | 13 | 2083 | |||
XM_017026744.1 | XP_016882233.1 | 12 | 2218 | |||
XM_017026745.1 | XP_016882234.1 | 12 | 2238 | |||
GenBank mRNA | AK131571.1 | - | 12 | 2206 |