Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001479.3 | NP_001001479.1 | 1 | 1046 | ||
NM_001318370.1 | NP_001305299.1 | 1 | 1046 | |||
NM_001318371.1 | NP_001305300.1 | 1 | 1046 | |||
XM_017028794.1 | XP_016884283.1 | 2 | 2138 | |||
XM_017028795.1 | XP_016884284.1 | 2 | 1712 | |||
GenBank mRNA | BC040191.1 | - | 1 | 1054 | 4814106 | |
BC093099.1 | - | 1 | 538 | 30511864 | ||
BC107119.2 | - | 1 | 432 | 40023535 | ||
CR456351.1 | - | 1 | 535 |