Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212555.2 | NP_997720.1 | 3, 4 | 259 | ||
GenBank mRNA | AY358105.1 | - | 3, 4 | 259 | ||
BC137190.1 | - | 3, 4 | 250 | 9021187 | ||
BC137193.1 | - | 3, 4 | 250 | 9021190 | ||
BC144526.1 | - | 2, 3 | 121 | 9053053 |