Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FBXL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099784.2 | NP_001093254.2 | 11 | 2721 | ||
NM_001282351.1 | NP_001269280.1 | 11 | 2589 | |||
GenBank mRNA | AK127701.1 | - | 11 | 2573 | ||
AL137589.1 | - | 3 | 931 | |||
BC059173.1 | - | 10 | 2237 | 5738074 |