Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099784.2 | NP_001093254.2 | 10, 11 | 2059 | ||
NM_001282351.1 | NP_001269280.1 | 10, 11 | 1927 | |||
GenBank mRNA | AK098777.1 | - | 8, 9 | 1588 | ||
AK127701.1 | - | 10, 11 | 1911 | |||
AK302669.1 | - | 10, 11 | 2061 | |||
AL137589.1 | - | 2, 3 | 269 | |||
BC059173.1 | - | 9, 10 | 1575 | 5738074 |