Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019107.3 | NP_061980.1 | 6 | 674 | ||
GenBank mRNA | AF282264.1 | - | 6 | 606 | ||
AK098829.1 | - | 6 | 613 | |||
AL365373.1 | - | 6 | 596 | |||
AL365374.1 | - | 6 | 596 | |||
BC003639.2 | - | 6 | 592 | |||
BC010129.2 | - | 6 | 641 | 3953027 | ||
BC014655.1 | - | 6 | 612 | 4562455 |