Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022149.4 | NP_071432.2 | 1 | 631 | ||
GenBank mRNA | AF295378.1 | - | 1 | 580 | ||
AF320910.1 | - | 1 | 499 | |||
BC010056.2 | - | 1 | 585 | 3943384 | ||
BQ058163.1 | - | 1 | 237 | |||
BQ070043.1 | - | 1 | 632 |