Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270891.1 | NP_001257820.1 | 1 | 42 | ||
NM_001270892.1 | NP_001257821.1 | 1 | 42 | |||
NM_001270893.1 | NP_001257822.1 | 1 | 42 | |||
NM_024108.2 | NP_077013.1 | 1 | 42 | |||
XM_017027295.1 | XP_016882784.1 | 1 | 101 | |||
GenBank mRNA | AF161407.1 | - | 1 | 36 | ||
BC001907.1 | - | 1 | 26 | 3534111 | ||
BC004450.1 | - | 1 | 27 | 2820830 | ||
BI462720.1 | - | 1 | 41 | |||
CN256686.1 | - | 1 | 37 | |||
CN256687.1 | - | 1 | 54 | |||
FJ418644.1 | - | 1 | 24 |