Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014661.3 | NP_055476.3 | 3 | 634 | ||
GenBank mRNA | AK127343.1 | - | 3 | 828 | ||
BC031654.1 | - | 3 | 542 | 5174078 | ||
BX344666.2 | - | 3 | 632 | |||
CA397342.1 | - | 3 | 547 | |||
D50930.1 | - | 3 | 323 |