Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032477.2 | NP_115866.1 | 2 | 477 | ||
GenBank mRNA | AI091177.1 | - | 2 | 477 | ||
AY232291.1 | - | 2 | 401 | |||
BC040035.1 | - | 2 | 452 | 5394644 | ||
BM919526.1 | - | 457 |