Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEPDC5 SIRNA Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5
Gene Aliases:
DEP.5, FFEVF
Chromosome Location:
Chr.22: 31753951 - 31907034 on Build GRCh38
UniGene ID:
Hs.435022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21574
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007188.2
NP_001007189.1
4
372
NM_001136029.2
NP_001129501.1
3
230
NM_001242896.1
NP_001229825.1
4
372
NM_001242897.1
NP_001229826.1
4
276
NM_014662.4
NP_055477.1
3
230
NR_110988.1
4
372
XM_005261862.1
XP_005261919.1
4
276
XM_011530557.2
XP_011528859.1
4
379
XM_011530558.2
XP_011528860.1
4
379
XM_011530559.2
XP_011528861.1
4
379
XM_011530560.2
XP_011528862.1
4
398
XM_011530561.2
XP_011528863.1
4
379
XM_011530562.2
XP_011528864.1
4
379
XM_011530563.2
XP_011528865.1
4
379
XM_011530565.2
XP_011528867.1
4
379
XM_011530568.2
XP_011528870.1
4
379
XM_017029109.1
XP_016884598.1
4
276
XM_017029110.1
XP_016884599.1
4
276
XM_017029111.1
XP_016884600.1
4
276
XM_017029112.1
XP_016884601.1
4
276
XM_017029113.1
XP_016884602.1
4
379
XM_017029114.1
XP_016884603.1
4
379
XM_017029115.1
XP_016884604.1
4
379
GenBank mRNA
AB014545.1
-
3
229
AJ698950.1
-
3
225
AJ698951.1
-
3
225
AJ704764.1
-
3
225
AK294987.1
-
4
248
AK300737.1
-
4
341
BC057797.1
-
4
312
BC136612.1
-
4
286
9020602
BC146766.1
-
3
229
9007209
BP354669.1
-
4
373
DA339804.1
-
4
276
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of TOR signaling
cellular response to amino acid starvation
intracellular signal transduction
positive regulation of GTPase activity
negative regulation of TORC1 signaling
GTPase activator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0