Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215790
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
18
3111
NM_001258250.1
NP_001245179.1
16
2400
NM_001258251.1
NP_001245180.1
17
2868
NM_001258252.1
NP_001245181.1
17
2955
NM_001258253.1
NP_001245182.1
17
2868
NM_001258254.1
NP_001245183.1
17
2583
NM_001258255.1
NP_001245184.1
17
2583
NM_001258256.1
NP_001245185.1
17
2955
NM_001258257.1
NP_001245186.1
16
2448
NM_001258258.1
NP_001245187.1
17
2976
NM_001258259.1
NP_001245188.1
16
2772
NM_001258260.1
NP_001245189.1
17
2910
NM_001258261.1
NP_001245190.1
17
2946
NM_001258262.1
NP_001245191.1
18
2739
NM_001258263.1
NP_001245192.1
16
2820
NM_001258264.1
NP_001245193.1
17
2907
NM_001258265.1
NP_001245194.1
12
2571
NM_001258266.1
NP_001245195.1
17
2955
NM_001258267.1
NP_001245196.1
16
2820
NM_001258268.1
NP_001245197.1
17
2583
NM_001258269.1
NP_001245198.1
18
2673
NM_001258270.1
NP_001245199.1
16
2448
NM_007125.4
NP_009056.3
16
2820
NM_182659.1
NP_872600.1
16
2820
NM_182660.1
NP_872601.1
16
2820
NR_047597.1
17
2926
NR_047598.1
18
3056
NR_047599.1
16
2820
NR_047600.1
17
2744
NR_047602.1
15
2785
NR_047603.1
17
2943
NR_047607.1
18
3166
NR_047609.1
16
2578
NR_047618.1
19
2829
NR_047624.1
18
3040
NR_047632.1
17
2528
NR_047635.1
16
2815
NR_047637.1
17
2878
NR_047638.1
16
2886
NR_047641.1
16
2820
NR_047642.1
16
2820
NR_047644.1
18
3013
NR_047645.1
17
3026
NR_047646.1
15
2693
NR_047647.1
16
2777
XM_005262518.3
XP_005262575.1
17
2301
XM_006724875.3
XP_006724938.1
18
2457
XM_011531441.2
XP_011529743.1
19
2505
XM_011531442.2
XP_011529744.1
18
2457
XM_011531443.2
XP_011529745.1
19
2439
XM_011531445.2
XP_011529747.1
18
2370
XM_011531446.2
XP_011529748.1
18
2349
XM_011531447.2
XP_011529749.1
17
2322
XM_011531448.2
XP_011529750.1
18
2304
XM_011531449.2
XP_011529751.1
17
2301
XM_011531450.2
XP_011529752.1
17
2256
XM_011531451.2
XP_011529753.1
17
2214
XM_011531452.2
XP_011529754.1
16
2166
XM_011531453.2
XP_011529755.1
19
2505
XM_011531454.2
XP_011529756.1
19
2505
XM_011531455.2
XP_011529757.1
19
2505
XM_011531457.2
XP_011529759.1
19
2505
XM_011531458.2
XP_011529760.1
18
2391
XM_011531459.2
XP_011529761.1
19
2505
XM_011531460.2
XP_011529762.1
19
2505
XM_011531461.2
XP_011529763.1
18
2457
XM_011531462.2
XP_011529764.1
18
2391
XM_011531463.2
XP_011529765.1
18
2349
XM_011531464.2
XP_011529766.1
17
2301
XM_011531465.2
XP_011529767.1
17
2214
XM_017030066.1
XP_016885555.1
18
2391
XM_017030067.1
XP_016885556.1
17
2301
XM_017030068.1
XP_016885557.1
16
2166
XM_017030069.1
XP_016885558.1
16
2166
XM_017030070.1
XP_016885559.1
19
2505
XM_017030071.1
XP_016885560.1
19
2505
XM_017030072.1
XP_016885561.1
18
2304
XM_017030073.1
XP_016885562.1
17
2214
XM_017030074.1
XP_016885563.1
19
2439
GenBank mRNA
AF000994.1
-
16
2820
AF000995.1
-
16
2820
AF000996.1
-
16
2820
AK292858.1
-
16
2070
AY682463.1
-
15
1722
AY730613.1
-
18
2019
AY769061.1
-
18
2051
AY769062.1
-
17
1921
AY769064.1
-
15
1780
AY769067.1
-
16
1815
AY769068.1
-
17
1873
AY934850.1
-
16
1395
AY934851.1
-
16
1767
AY934852.1
-
16
1806
AY934853.1
-
16
1796
AY934855.1
-
15
1716
AY934856.1
-
15
1732
AY934857.1
-
17
1929
CR936684.1
-
8
918
DQ012094.1
-
5
420
DQ140389.1
-
17
1950
DQ140390.1
-
17
1578
DQ341385.1
-
17
1863
DQ341386.1
-
16
1815
DQ341387.1
-
16
1764
DQ345543.1
-
17
1905
DQ345544.1
-
18
2040
DQ350456.1
-
17
1739
DQ350457.1
-
17
1863
DQ350459.1
-
16
1815
EF612660.1
-
17
1578
EF612661.1
-
15
1716
EF612662.1
-
17
1578
EF612663.1
-
17
1950
EF612664.1
-
16
1443
EF612665.1
-
16
1767
EF612667.1
-
15
1716
EF612668.1
-
18
2106
EF612669.1
-
16
1443
EF612670.1
-
15
1671
EF612671.1
-
15
1344
EF612672.1
-
16
1767
EF612673.1
-
17
1971
EF612674.1
-
16
1767
EF612675.1
-
17
1902
EF612676.1
-
16
1443
EF612677.1
-
18
2106
EF612678.1
-
17
1491
EF612679.1
-
18
1668
EF612680.1
-
17
1905
EF612681.1
-
14
1353
EF612684.1
-
18
1734
EF612685.1
-
18
2040
EF612686.1
-
17
1950
EF612687.1
-
16
1863
EF612688.1
-
16
1395
EF612689.1
-
18
1998
EF612690.1
-
16
1764
EF612691.1
-
17
1950
EF612692.1
-
14
1725
EF612693.1
-
16
1815
EF612694.1
-
17
1491
EF612695.1
-
17
1902
EF612696.1
-
17
1905
EF612697.1
-
17
1863
EF612698.1
-
14
1725
EF612699.1
-
16
1443
EF653165.1
-
15
1780
EF653169.1
-
15
1780
EF653170.1
-
14
1681
EF653171.1
-
17
1863
EF653172.1
-
15
1706
EF653173.1
-
16
1815
EF653175.1
-
18
2056
EF653177.1
-
18
1653
EF653178.1
-
19
2172
EF653181.1
-
18
1953
EF653182.1
-
15
1780
EF653183.1
-
17
1921
EF653184.1
-
17
1929
EF653188.1
-
12
1017
EF653189.1
-
12
1566
EF653190.1
-
17
1950
EF653196.1
-
16
1815
EF653198.1
-
16
1815
EF653199.1
-
18
2035
EF653201.1
-
16
1856
EF653202.1
-
14
1594
EF653203.1
-
15
1780
EF653204.1
-
18
1979
EF653205.1
-
17
1863
EF653207.1
-
18
2008
EF653208.1
-
17
1921
EF653209.1
-
17
2021
EF653210.1
-
3
261
EF653211.1
-
18
1979
EF653213.1
-
15
1688
EF653214.1
-
19
2180
EF653216.1
-
16
1867
EF653217.1
-
16
1772
EF653219.1
-
18
2106
EF653220.1
-
18
1734
EF653222.1
-
19
1814
EF653224.1
-
18
2161
EF685284.1
-
16
1573
EF685303.1
-
16
1815
EF685307.1
-
18
2035
EF685315.1
-
15
1721
EF685323.1
-
15
1716
EF685324.1
-
15
1632
EF685325.1
-
17
1841
EF685326.1
-
17
1873
EF685327.1
-
17
2059
EF685332.1
-
17
1950
EF685333.1
-
18
1941
EF685334.1
-
16
1815
EF685336.1
-
16
1443
EF685338.1
-
16
1645
EF685339.1
-
15
1716
EF685341.1
-
16
1764
EU026275.1
-
16
1822
EU026277.1
-
17
1881
EU026280.1
-
19
2079
EU026281.1
-
15
1780
EU026289.1
-
15
1939
EU026292.1
-
20
1804
EU026300.1
-
16
1828
EU026303.1
-
17
1945
EU026306.1
-
17
1929
EU026308.1
-
15
1787
EU026314.1
-
18
2056
EU026316.1
-
17
1523
EU026318.1
-
14
1682
EU026322.1
-
17
1858
EU026323.1
-
16
1910
EU026324.1
-
14
1605
EU026334.1
-
15
1713
EU026335.1
-
18
2008
EU026339.1
-
14
1681
EU026341.1
-
9
1214
EU026344.1
-
16
1780
EU026345.1
-
16
1810
EU026347.1
-
14
1681
EU026350.1
-
16
1759
EU116344.1
-
7
788
EU116346.1
-
16
1815
EU116347.1
-
15
1821
EU116350.1
-
16
1743
EU116351.1
-
11
1327
EU116352.1
-
16
1810
EU163941.1
-
16
1881
EU163942.1
-
17
1873
EU163943.1
-
7
1038
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0