Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288781.1 | NP_001275710.1 | 16 | 1929 | ||
NM_001288782.1 | NP_001275711.1 | 14 | 1849 | |||
NM_001288783.1 | NP_001275712.1 | 15 | 1821 | |||
NM_144596.3 | NP_653197.2 | 16 | 1881 | |||
NM_198309.3 | NP_938051.1 | 15 | 1851 | |||
NM_198310.3 | NP_938052.1 | 14 | 1761 | |||
XM_006720035.1 | XP_006720098.1 | 14 | 2133 | |||
XM_006720037.2 | XP_006720100.1 | 13 | 2043 | |||
XM_011536432.1 | XP_011534734.1 | 15 | 2211 | |||
XM_011536433.1 | XP_011534735.1 | 14 | 2127 | |||
XM_011536434.1 | XP_011534736.1 | 14 | 2121 | |||
XM_011536435.2 | XP_011534737.1 | 14 | 1597 | |||
XM_017020977.1 | XP_016876466.1 | 13 | 2049 | |||
XM_017020978.1 | XP_016876467.1 | 12 | 1959 | |||
GenBank mRNA | AF086168.1 | - | 2 | 276 | ||
AK093891.1 | - | 14 | 1699 | |||
AK124675.1 | - | 15 | 1731 | |||
AL833901.1 | - | 13 | 1585 | |||
AY366523.1 | - | 14 | 1707 | |||
AY366524.1 | - | 13 | 1617 | |||
AY373972.1 | - | 14 | 1707 | |||
BC001563.1 | - | 10 | 1199 | |||
BC026351.1 | - | 15 | 1785 | 4799330 | ||
BC095433.1 | - | 14 | 1689 | 30368216 | ||
BX161472.1 | - | 13 | 1689 | |||
BX247959.1 | - | 14 | 1798 | |||
BX248071.1 | - | 6 | 938 | |||
BX248248.1 | - | 11 | 1293 | |||
BX537546.1 | - | 1 | 3609 |