Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DALRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009996.2 | NP_001009996.1 | 9 | 1254 | ||
NM_001276405.1 | NP_001263334.1 | 9 | 1254 | |||
NM_018114.5 | NP_060584.3 | 9 | 1271 | |||
XM_005265269.3 | XP_005265326.1 | 9 | 1235 | |||
XM_006713219.2 | XP_006713282.1 | 10 | 1390 | |||
XM_011533891.1 | XP_011532193.1 | 9 | 1235 | |||
XM_011533892.1 | XP_011532194.1 | 9 | 1586 | |||
XM_011533893.1 | XP_011532195.1 | 8 | 1173 | |||
XM_017006723.1 | XP_016862212.1 | 10 | 1390 | |||
GenBank mRNA | AK001358.1 | - | 8 | 1239 | ||
AK093054.1 | - | 2 | 2201 | |||
AK093204.1 | - | 9 | 1235 | |||
AK093294.1 | - | 8 | 1042 | |||
AK301352.1 | - | 9 | 1222 | |||
AK308273.1 | - | 10 | 1348 | |||
BC014099.2 | - | 8 | 1293 | |||
BC032440.1 | - | 9 | 1217 | 5163675 | ||
BC047683.1 | - | 9 | 1216 | 5755743 | ||
BC054493.1 | - | 9 | 1210 | 5755664 | ||
BX648719.1 | - | 6 | 839 |