Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018405.3 | NP_060875.2 | 4 | 710 | ||
XM_005258000.1 | XP_005258057.1 | 4 | 820 | |||
GenBank mRNA | AJ272196.2 | - | 4 | 617 | ||
AK024772.1 | - | 4 | 630 | |||
BC012386.1 | - | 4 | 640 | 3882737 | ||
BC035483.1 | - | 5 | 842 | |||
BQ671018.1 | - | 1 | 200 |